siRNA Details
| Gene Symbol: Spink2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008870.2 | NP_001008870.1 | 3, 4 | 222 | ||
| GenBank mRNA | CR477121.1 | - | 1, 2 | 287 | ||
| Gene Symbol: LOC100910138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751350.3 | XP_003751398.1 | 3, 4 | 327 | ||

Spink2
Genomic Map