siRNA Details
| Gene Symbol: Prss57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003956.1 | NP_001003956.1 | 2 | 105 | ||
| XM_006240979.2 | XP_006241041.1 | 2 | 248 | |||
| XM_008765175.1 | XP_008763397.1 | 2 | 248 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss57| Gene Symbol: Prss57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003956.1 | NP_001003956.1 | 2 | 105 | ||
| XM_006240979.2 | XP_006241041.1 | 2 | 248 | |||
| XM_008765175.1 | XP_008763397.1 | 2 | 248 | |||