siRNA Details
| Gene Symbol: Prss45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008864.1 | NP_001008864.1 | 6 | 813 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss45| Gene Symbol: Prss45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008864.1 | NP_001008864.1 | 6 | 813 | ||