siRNA Details
| Gene Symbol: RT1-M1-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008849.1 | NP_001008849.1 | 2 | 173 | ||
| XM_006255884.2 | XP_006255946.1 | 3 | 595 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RT1-M1-2| Gene Symbol: RT1-M1-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008849.1 | NP_001008849.1 | 2 | 173 | ||
| XM_006255884.2 | XP_006255946.1 | 3 | 595 | |||