siRNA Details
| Gene Symbol: Gpank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034157.2 | NP_001029329.1 | 3 | 1055 | ||
| XM_006256075.2 | XP_006256137.1 | 3 | 972 | |||
| XM_006256076.2 | XP_006256138.1 | 3 | 1126 | |||
| XM_006256077.2 | XP_006256139.1 | 4 | 1379 | |||
| XM_008772761.1 | XP_008770983.1 | 3 | 1096 | |||
| GenBank mRNA | BU758690.1 | - | 1 | 367 | ||
| FQ213768.1 | - | 3 | 1014 | |||

Gpank1
Genomic Map