siRNA Details
| Gene Symbol: Npc1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002025.1 | NP_001002025.1 | 5 | 1910 | ||
| GenBank mRNA | AY437867.1 | - | 5 | 1910 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npc1l1| Gene Symbol: Npc1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002025.1 | NP_001002025.1 | 5 | 1910 | ||
| GenBank mRNA | AY437867.1 | - | 5 | 1910 | ||