siRNA Details
| Gene Symbol: Atp6v0e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002253.1 | NP_001002253.1 | 2 | 193 | ||
| GenBank mRNA | BC089958.1 | - | 145 | 7312897 | ||
| CB757317.1 | - | 2 | 177 | |||
| FQ212114.1 | - | 1283 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp6v0e2| Gene Symbol: Atp6v0e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002253.1 | NP_001002253.1 | 2 | 193 | ||
| GenBank mRNA | BC089958.1 | - | 145 | 7312897 | ||
| CB757317.1 | - | 2 | 177 | |||
| FQ212114.1 | - | 1283 | ||||