siRNA Details
| Gene Symbol: Atp6v0e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002253.1 | NP_001002253.1 | 1 | 90 | ||
| GenBank mRNA | BC089958.1 | - | 42 | 7312897 | ||
| CB566985.1 | - | 1 | 113 | |||
| CB757317.1 | - | 1 | 74 | |||
| FQ212114.1 | - | 1180 | ||||

Atp6v0e2
Genomic Map