siRNA Details
| Gene Symbol: Ufc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003709.2 | NP_001003709.1 | 1 | 289 | ||
| XM_006250251.2 | XP_006250313.1 | 2 | 244 | |||
| GenBank mRNA | AB186051.1 | - | 1 | 207 | ||
| AB441169.1 | - | 1 | 207 | |||
| BC087648.1 | - | 1 | 289 | 7310651 | ||
| FQ212748.1 | - | 1 | 307 | |||
| FQ213023.1 | - | 1 | 280 | |||
| FQ213507.1 | - | 1 | 290 | |||
| FQ216057.1 | - | 1 | 284 | |||
| FQ218926.1 | - | 1 | 310 | |||
| FQ219875.1 | - | 1 | 159 | |||
| FQ226625.1 | - | 1 | 302 | |||
| FQ232457.1 | - | 1 | 266 | |||

Ufc1
Genomic Map