siRNA Details
Gene Symbol: Ufc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003709.2 | NP_001003709.1 | 3, 4 | 458 | ||
XM_006250251.2 | XP_006250313.1 | 4, 5 | 413 | |||
GenBank mRNA | AB186051.1 | - | 3, 4 | 376 | ||
BC087648.1 | - | 3, 4 | 458 | 7310651 | ||
FQ212748.1 | - | 3, 4 | 476 | |||
FQ213023.1 | - | 3, 4 | 449 | |||
FQ213507.1 | - | 3, 4 | 459 | |||
FQ216057.1 | - | 3, 4 | 453 | |||
FQ218926.1 | - | 3, 4 | 479 | |||
FQ219875.1 | - | 3, 4 | 328 | |||
FQ226625.1 | - | 3, 4 | 471 |
