siRNA Details
| Gene Symbol: Ufc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003709.2 | NP_001003709.1 | 6 | 658 | ||
| XM_006250251.2 | XP_006250313.1 | 7 | 613 | |||
| GenBank mRNA | AB186051.1 | - | 6 | 576 | ||
| AB441169.1 | - | 6 | 605 | |||
| BC087648.1 | - | 6 | 658 | 7310651 | ||
| FQ212748.1 | - | 6 | 676 | |||
| FQ213507.1 | - | 6 | 659 | |||
| FQ216057.1 | - | 6 | 653 | |||
| FQ218926.1 | - | 6 | 679 | |||
| FQ219875.1 | - | 6 | 528 | |||
| FQ232457.1 | - | 6 | 635 | |||

Ufc1
Genomic Map