siRNA Details
| Gene Symbol: Jph4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003711.1 | NP_001003711.1 | 2, 3 | 1147 | ||
| XM_008770690.1 | XP_008768912.1 | 3, 4 | 1945 | |||
| GenBank mRNA | AY341260.1 | - | 2, 3 | 1147 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jph4| Gene Symbol: Jph4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003711.1 | NP_001003711.1 | 2, 3 | 1147 | ||
| XM_008770690.1 | XP_008768912.1 | 3, 4 | 1945 | |||
| GenBank mRNA | AY341260.1 | - | 2, 3 | 1147 | ||