siRNA Details
| Gene Symbol: Smgc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004449.1 | NP_001004449.1 | 16 | 1795 | ||
| GenBank mRNA | AY459347.1 | - | 16 | 1795 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smgc| Gene Symbol: Smgc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004449.1 | NP_001004449.1 | 16 | 1795 | ||
| GenBank mRNA | AY459347.1 | - | 16 | 1795 | ||