siRNA Details
| Gene Symbol: Clec4e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005897.1 | NP_001005897.1 | 4 | 354 | ||
| GenBank mRNA | AY363175.1 | - | 4 | 354 | ||
| AY494064.1 | - | 4 | 320 | |||
| AY581049.1 | - | 4 | 320 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec4e| Gene Symbol: Clec4e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005897.1 | NP_001005897.1 | 4 | 354 | ||
| GenBank mRNA | AY363175.1 | - | 4 | 354 | ||
| AY494064.1 | - | 4 | 320 | |||
| AY581049.1 | - | 4 | 320 | |||