siRNA Details
| Gene Symbol: Krt27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008824.1 | NP_001008824.1 | 3 | 621 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt27| Gene Symbol: Krt27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008824.1 | NP_001008824.1 | 3 | 621 | ||