siRNA Details
| Gene Symbol: Krt32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008818.1 | NP_001008818.1 | 2 | 510 | ||
| XM_006247394.2 | XP_006247456.1 | 2 | 534 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt32| Gene Symbol: Krt32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008818.1 | NP_001008818.1 | 2 | 510 | ||
| XM_006247394.2 | XP_006247456.1 | 2 | 534 | |||