siRNA Details
| Gene Symbol: Hcst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005900.1 | NP_001005900.1 | 3 | 239 | ||
| XM_006228785.2 | XP_006228847.1 | 3 | 503 | |||
| GenBank mRNA | AY247020.1 | - | 3 | 239 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hcst| Gene Symbol: Hcst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005900.1 | NP_001005900.1 | 3 | 239 | ||
| XM_006228785.2 | XP_006228847.1 | 3 | 503 | |||
| GenBank mRNA | AY247020.1 | - | 3 | 239 | ||