siRNA Details
| Gene Symbol: Hcst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005900.1 | NP_001005900.1 | 3, 4 | 280 | ||
| GenBank mRNA | AY247020.1 | - | 3, 4 | 280 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hcst| Gene Symbol: Hcst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005900.1 | NP_001005900.1 | 3, 4 | 280 | ||
| GenBank mRNA | AY247020.1 | - | 3, 4 | 280 | ||