siRNA Details
| Gene Symbol: Ispd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008386.1 | NP_001008387.1 | 8 | 1136 | ||
| GenBank mRNA | AY769991.1 | - | 7 | 821 | ||
| AY769992.1 | - | 8 | 1136 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ispd| Gene Symbol: Ispd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008386.1 | NP_001008387.1 | 8 | 1136 | ||
| GenBank mRNA | AY769991.1 | - | 7 | 821 | ||
| AY769992.1 | - | 8 | 1136 | |||