siRNA Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008527.1 | NP_001008527.1 | 12, 13 | 1218 | ||
| XM_006245004.2 | XP_006245066.1 | 12, 13 | 1246 | |||
| XM_008767121.1 | XP_008765343.1 | 12, 13 | 1247 | |||
| XM_008767122.1 | XP_008765344.1 | 8, 9 | 1915 | |||
| XM_008767123.1 | XP_008765345.1 | 6, 7 | 737 | |||
| XM_008767124.1 | XP_008765346.1 | 7, 8 | 913 | |||
| GenBank mRNA | AY665586.1 | - | 12, 13 | 1218 | ||

Aox3
Genomic Map