siRNA Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008527.1 | NP_001008527.1 | 33, 34 | 3870 | ||
| XM_006245004.2 | XP_006245066.1 | 33, 34 | 3898 | |||
| XM_008767121.1 | XP_008765343.1 | 33, 34 | 3899 | |||
| XM_008767122.1 | XP_008765344.1 | 29, 30 | 4567 | |||
| XM_008767123.1 | XP_008765345.1 | 27, 28 | 3389 | |||
| XM_008767124.1 | XP_008765346.1 | 28, 29 | 3565 | |||
| GenBank mRNA | AY665586.1 | - | 33, 34 | 3870 | ||
| Gene Symbol: Aox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY325166.1 | - | 98 | |||

Aox3
Genomic Map