siRNA Details
| Gene Symbol: Klra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009718.1 | NP_001009718.1 | 4 | 460 | ||
| XM_008763403.1 | XP_008761625.1 | 3 | 522 | |||
| GenBank mRNA | AY651018.1 | - | 4 | 460 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klra2| Gene Symbol: Klra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009718.1 | NP_001009718.1 | 4 | 460 | ||
| XM_008763403.1 | XP_008761625.1 | 3 | 522 | |||
| GenBank mRNA | AY651018.1 | - | 4 | 460 | ||