siRNA Details
Gene Symbol: Oas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009493.1 | NP_001009493.1 | 2177 | |||
GenBank mRNA | AY250706.1 | - | 2177 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Oas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009493.1 | NP_001009493.1 | 2177 | |||
GenBank mRNA | AY250706.1 | - | 2177 |