siRNA Details
| Gene Symbol: Vom1r4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009516.1 | NP_001009516.1 | 1 | 541 | ||
| GenBank mRNA | AY510338.1 | - | 1 | 541 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r4| Gene Symbol: Vom1r4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009516.1 | NP_001009516.1 | 1 | 541 | ||
| GenBank mRNA | AY510338.1 | - | 1 | 541 | ||