siRNA Details
Gene Symbol: Vom1r70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009519.1 | NP_001009519.1 | 1 | 130 | ||
GenBank mRNA | AY510318.1 | - | 1 | 130 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vom1r70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009519.1 | NP_001009519.1 | 1 | 130 | ||
GenBank mRNA | AY510318.1 | - | 1 | 130 |