siRNA Details
| Gene Symbol: Vom1r76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009520.1 | NP_001009520.1 | 1 | 166 | ||
| GenBank mRNA | AY510327.1 | - | 1 | 166 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r76| Gene Symbol: Vom1r76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009520.1 | NP_001009520.1 | 1 | 166 | ||
| GenBank mRNA | AY510327.1 | - | 1 | 166 | ||