siRNA Details
| Gene Symbol: Vom1r44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008902.1 | NP_001008902.1 | 1 | 721 | ||
| XM_008758769.1 | XP_008756991.1 | 3 | 1743 | |||
| XM_008758770.1 | XP_008756992.1 | 4 | 1265 | |||
| GenBank mRNA | AY510307.1 | - | 1 | 721 | ||

Vom1r44
Genomic Map