siRNA Details
Gene Symbol: Vom1r89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008928.1 | NP_001008928.1 | 1 | 848 | ||
GenBank mRNA | AY510298.1 | - | 1 | 848 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vom1r89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008928.1 | NP_001008928.1 | 1 | 848 | ||
GenBank mRNA | AY510298.1 | - | 1 | 848 |