siRNA Details
| Gene Symbol: Vom1r48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008931.1 | NP_001008931.1 | 19 | |||
| GenBank mRNA | AY510315.1 | - | 19 | |||
| Gene Symbol: LOC103691006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758859.1 | XP_008757081.1 | 1 | 36 | ||

Vom1r48
Genomic Map