siRNA Details
| Gene Symbol: Vom1r23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008941.1 | NP_001008941.1 | 1 | 875 | ||
| GenBank mRNA | AY510359.1 | - | 1 | 875 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r23| Gene Symbol: Vom1r23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008941.1 | NP_001008941.1 | 1 | 875 | ||
| GenBank mRNA | AY510359.1 | - | 1 | 875 | ||