siRNA Details
| Gene Symbol: Vom1r11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008955.1 | NP_001008955.1 | 1 | 383 | ||
| GenBank mRNA | AY510291.1 | - | 1 | 383 | ||
| Gene Symbol: LOC103690997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758821.1 | XP_008757043.1 | 410 | |||

Vom1r11
Genomic Map