siRNA Details
| Gene Symbol: Zfp868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009538.1 | NP_001009538.1 | 5 | 1450 | ||
| XM_006252915.2 | XP_006252977.1 | 6 | 1607 | |||
| XM_006252916.2 | XP_006252978.1 | 6 | 1818 | |||
| XM_006252917.2 | XP_006252979.1 | 5 | 1743 | |||
| GenBank mRNA | BC062078.1 | - | 5 | 1450 | 5599629 | |
| BC089094.1 | - | 5 | 1473 | 7125789 | ||

Zfp868
Genomic Map