siRNA Details
| Gene Symbol: Tacstd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009540.2 | NP_001009540.2 | 1 | 878 | ||
| GenBank mRNA | AY185508.1 | - | 1 | 878 | ||
| BC060519.1 | - | 1 | 859 | 5597863 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tacstd2| Gene Symbol: Tacstd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009540.2 | NP_001009540.2 | 1 | 878 | ||
| GenBank mRNA | AY185508.1 | - | 1 | 878 | ||
| BC060519.1 | - | 1 | 859 | 5597863 | ||