siRNA Details
| Gene Symbol: Cdkn2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009719.1 | NP_001009719.1 | 455 | |||
| GenBank mRNA | BC063185.1 | - | 455 | 6889848 | ||
| BC088350.1 | - | 427 | 7368483 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdkn2d| Gene Symbol: Cdkn2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009719.1 | NP_001009719.1 | 455 | |||
| GenBank mRNA | BC063185.1 | - | 455 | 6889848 | ||
| BC088350.1 | - | 427 | 7368483 | |||