siRNA Details
Gene Symbol: Ccdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011561.1 | NP_001011561.1 | 15 | 1685 | ||
GenBank mRNA | BC079021.1 | - | 15 | 1685 | 7113858 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011561.1 | NP_001011561.1 | 15 | 1685 | ||
GenBank mRNA | BC079021.1 | - | 15 | 1685 | 7113858 |