siRNA Details
| Gene Symbol: Xkrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012230.1 | NP_001012230.1 | 3 | 1364 | ||
| GenBank mRNA | AY534257.1 | - | 3 | 1364 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xkrx| Gene Symbol: Xkrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012230.1 | NP_001012230.1 | 3 | 1364 | ||
| GenBank mRNA | AY534257.1 | - | 3 | 1364 | ||