siRNA Details
| Gene Symbol: Plcz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012234.1 | NP_001012234.1 | 1, 2 | 26 | ||
| XM_006237597.2 | XP_006237659.1 | 2, 3 | 255 | |||
| GenBank mRNA | AY885259.1 | - | 1, 2 | 26 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plcz1| Gene Symbol: Plcz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012234.1 | NP_001012234.1 | 1, 2 | 26 | ||
| XM_006237597.2 | XP_006237659.1 | 2, 3 | 255 | |||
| GenBank mRNA | AY885259.1 | - | 1, 2 | 26 | ||