siRNA Details
Gene Symbol: Fut10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012360.1 | NP_001012360.1 | 3 | 1285 | ||
GenBank mRNA | AJ879584.1 | - | 3 | 1285 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fut10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012360.1 | NP_001012360.1 | 3 | 1285 | ||
GenBank mRNA | AJ879584.1 | - | 3 | 1285 |