siRNA Details
| Gene Symbol: Fut10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012360.1 | NP_001012360.1 | 2 | 177 | ||
| XM_006253236.2 | XP_006253298.1 | 3 | 562 | |||
| GenBank mRNA | AJ879584.1 | - | 2 | 177 | ||
| AJ880010.1 | - | 2 | 177 | |||

Fut10
Genomic Map