siRNA Details
| Gene Symbol: Cd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012830.1 | NP_036962.1 | 2 | 332 | ||
| GenBank mRNA | BC088164.1 | - | 2 | 262 | 7374969 | |
| X05111.1 | - | 2 | 332 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd2| Gene Symbol: Cd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012830.1 | NP_036962.1 | 2 | 332 | ||
| GenBank mRNA | BC088164.1 | - | 2 | 262 | 7374969 | |
| X05111.1 | - | 2 | 332 | |||