siRNA Details
| Gene Symbol: Scn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013119.1 | NP_037251.1 | 21, 22 | 4239 | ||
| GenBank mRNA | Y00766.1 | - | 21, 22 | 4239 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scn3a| Gene Symbol: Scn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013119.1 | NP_037251.1 | 21, 22 | 4239 | ||
| GenBank mRNA | Y00766.1 | - | 21, 22 | 4239 | ||