siRNA Details
Gene Symbol: Cps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017072.1 | NP_058768.1 | 25, 26 | 3149 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017072.1 | NP_058768.1 | 25, 26 | 3149 |