siRNA Details
| Gene Symbol: Mrpl28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_573060.5 | XP_573060.3 | 2 | 487 | ||
| GenBank mRNA | FQ219872.1 | - | 2 | 459 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Mrpl28
Mrpl28| Gene Symbol: Mrpl28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_573060.5 | XP_573060.3 | 2 | 487 | ||
| GenBank mRNA | FQ219872.1 | - | 2 | 459 | ||
 Back To Top
 Back To Top