siRNA Details
| Gene Symbol: Zfp62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246188.2 | XP_006246250.1 | 3 | 2750 | ||
| XM_006246189.2 | XP_006246251.1 | 2 | 2697 | |||
| XM_006246193.2 | XP_006246255.1 | 3 | 2746 | |||
| XM_008767726.1 | XP_008765948.1 | 2 | 6234 | |||
| XM_008767727.1 | XP_008765949.1 | 2 | 6230 | |||

Zfp62
Genomic Map