siRNA Details
| Gene Symbol: RGD1564036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109030.2 | NP_001102500.2 | 7 | 993 | ||
| XM_008767694.1 | XP_008765916.1 | 7 | 1567 | |||
| GenBank mRNA | BC166832.1 | - | 7 | 993 | 8375915 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564036| Gene Symbol: RGD1564036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109030.2 | NP_001102500.2 | 7 | 993 | ||
| XM_008767694.1 | XP_008765916.1 | 7 | 1567 | |||
| GenBank mRNA | BC166832.1 | - | 7 | 993 | 8375915 | |