siRNA Details
| Gene Symbol: Zfp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127558.1 | NP_001121030.1 | 5 | 1218 | ||
| XM_008767695.1 | XP_008765917.1 | 6 | 1282 | |||
| XM_008767697.1 | XP_008765919.1 | 6 | 1290 | |||
| XM_008767698.1 | XP_008765920.1 | 6 | 1243 | |||
| XM_008767699.1 | XP_008765921.1 | 6 | 1246 | |||
| GenBank mRNA | BC161925.1 | - | 5 | 1218 | 8372872 | |

Zfp2
Genomic Map