siRNA Details
| Gene Symbol: Ccdc69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109031.1 | NP_001102501.1 | 4 | 353 | ||
| XM_006246393.2 | XP_006246455.1 | 5 | 459 | |||
| XM_006246394.2 | XP_006246456.1 | 4 | 574 | |||
| XM_006246395.2 | XP_006246457.1 | 4 | 572 | |||
| XM_008767703.1 | XP_008765925.1 | 4 | 683 | |||

Ccdc69
Genomic Map