siRNA Details
| Gene Symbol: RGD1559732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750838.3 | XP_003750886.1 | 7 | 1410 | ||
| XM_006246534.2 | XP_006246596.1 | 6 | 1335 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1559732| Gene Symbol: RGD1559732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750838.3 | XP_003750886.1 | 7 | 1410 | ||
| XM_006246534.2 | XP_006246596.1 | 6 | 1335 | |||