siRNA Details
Gene Symbol: Pik3r5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191923.1 | NP_001178852.1 | 14 | 2163 | ||
XM_006246770.2 | XP_006246832.1 | 15 | 2308 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pik3r5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191923.1 | NP_001178852.1 | 14 | 2163 | ||
XM_006246770.2 | XP_006246832.1 | 15 | 2308 |