siRNA Details
| Gene Symbol: LOC497934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017474.1 | NP_001017474.2 | 1 | 972 | ||
| GenBank mRNA | BC082026.1 | - | 1 | 972 | 7129979 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC497934| Gene Symbol: LOC497934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017474.1 | NP_001017474.2 | 1 | 972 | ||
| GenBank mRNA | BC082026.1 | - | 1 | 972 | 7129979 | |